Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-phenylnaphthalene 98.0+%, TCI America™
SDP

Catalog No. B46361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46361G 1 g
B4636200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46361G Supplier TCI America Supplier No. B46361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22082-93-5
Molecular Formula C16H11Br
Molecular Weight (g/mol) 283.168
InChI Key ANKXTSUKPPHQJR-UHFFFAOYSA-N
PubChem CID 10956972
IUPAC Name 1-bromo-2-phenylnaphthalene
SMILES C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C=C2)Br

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 170°C
Quantity 1 g
Formula Weight 283.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-phenylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.