Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,3-dimethyl-2-butanone 95.0+%, TCI America™
SDP

Catalog No. B149025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B149025G 25 g
B14905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B149025G Supplier TCI America Supplier No. B149025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5469-26-1
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
MDL Number MFCD00000206
InChI Key SAIRZMWXVJEBMO-UHFFFAOYSA-N
Synonym 1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
PubChem CID 21642
IUPAC Name 1-bromo-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CBr

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 g
Formula Weight 179.06
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,3-dimethyl-2-butanone
RTECSNumber EL7010000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.