Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,4-difluorobenzene, 98%, Thermo Scientific™

Catalog No. AC233190010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-61-8
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00000304
InChI Key YMQPKONILWWJQG-UHFFFAOYSA-N
Synonym 1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
PubChem CID 67675
IUPAC Name 4-bromo-1,2-difluorobenzene
SMILES FC1=CC=C(Br)C=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -4°C
Color Colorless to Yellow
Density 1.7070g/mL
Boiling Point 150°C to 151°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.506
Linear Formula BrC6H3F2
Quantity 1 g
Specific Gravity 1.707
Solubility Information Solubility in water: insoluble
Formula Weight 193
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3, 4-difluorobenzene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-481-5
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.