Learn More
1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%
Description
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | -16°C |
Density | 1.698 |
Boiling Point | 152°C to 155°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.427 |
Quantity | 50 g |
Beilstein | 2123669 |
Solubility Information | Immiscible with water. |
Formula Weight | 293.01 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 206-334-5 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.