Learn More
1-Bromo-3,5-dimethoxybenzene, 97%
Description
1-Bromo-3,5-dimethoxybenzene is used as an intermediate in the synthetic preparation of pharmaceutical inhibitors via cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromo-3,5-dimethoxybenzene is used as an intermediate in the synthetic preparation of pharmaceutical inhibitors via cross-coupling reactions.
Solubility
Soluble in methanol.
Notes
Reacts with strong oxidizing agents.
            Specifications
Specifications
| Melting Point | 62°C to 66°C | 
| Quantity | 5 g | 
| Solubility Information | Soluble in methanol. | 
| Formula Weight | 217.06 | 
| Percent Purity | 97% | 
| Chemical Name or Material | 1-Bromo-3,5-dimethoxybenzene | 
Safety and Handling
| Hazard Category | H315-H319-H335 | 
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c | 
| TSCA | No | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.