Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,5-dimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog No. p-4258797
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20469-65-2
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD06200685
InChI Key KRWRFIMBWRVMKE-UHFFFAOYSA-N
Synonym 3,5-dimethoxybromobenzene, benzene, 1-bromo-3,5-dimethoxy, 1,3-dimethoxy-5-bromobenzene, 1-bromo-3,5-dimethoxy-benzene, 1-bromo-3,5-dimethox, 3,5-dimethoxy-bromobenzene, bromo-3,5-dimethoxybenzene, 3,5-dimethoxy bromobenzene, acmc-1cs59, ksc495e7l
PubChem CID 639187
IUPAC Name 1-bromo-3,5-dimethoxybenzene
SMILES COC1=CC(OC)=CC(Br)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63.6°C to 65.7°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 217.06
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-Bromo-3, 5-dimethoxybenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.