Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,5-dimethyladamantane 98.0+%, TCI America™
SDP

Catalog No. B182125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B182125G 25 g
B18215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B182125G Supplier TCI America Supplier No. B182125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 941-37-7
Molecular Formula C12H19Br
Molecular Weight (g/mol) 243.188
MDL Number MFCD00077197
InChI Key QUCXLVDIVQWYJR-UHFFFAOYSA-N
Synonym 1-bromo-3,5-dimethyl-adamantane, tricyclo 3.3.1.13,7 decane, 1-bromo-3,5-dimethyl, acmc-209rqd, 3,5-dimethyladamantylbromide, 1-bromo-3,5-dimethyltricyclo 3.3.1.13,7 decane, 1-bromo-3,5dimethyladamantane, 3,5-dimethyl-1-bromoadamantane, 1-bromo-3,5-dimethyl adamantane, 1-bromanyl-3,5-dimethyl-adamantane, 1-?bromo-?3,5-?dimethyladamantane
PubChem CID 98317
IUPAC Name 1-bromo-3,5-dimethyladamantane
SMILES CC12CC3CC(C1)(CC(C3)(C2)Br)C

Specifications

Color Yellow
Boiling Point 273°C
Quantity 25 g
Formula Weight 243.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,5-dimethyladamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.