Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-phenylimidazo[1,5-a]pyridine 97.0+%, TCI America™
SDP

Catalog No. B34385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34385G 5 g
B34381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34385G Supplier TCI America Supplier No. B34385G

Chemical Identifiers

CAS 104202-15-5
Molecular Formula C13H9BrN2
Molecular Weight (g/mol) 273.13
MDL Number MFCD04587511
InChI Key ULKZZBGVTIZXSA-UHFFFAOYSA-N
PubChem CID 957540
IUPAC Name 1-bromo-3-phenylimidazo[1,5-a]pyridine
SMILES BrC1=C2C=CC=CN2C(=N1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 273.13
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3-phenylimidazo[1,5-a]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.