Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-chlorobenzene, 98+%

Catalog No. AAA121600B
Click to view available options
Quantity:
100 g
1000 g

CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol

1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

Solubility
Soluble in water 0.045g/L.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-39-8
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000600
InChI Key NHDODQWIKUYWMW-UHFFFAOYSA-N
Synonym 4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
PubChem CID 7806
IUPAC Name 1-bromo-4-chlorobenzene
SMILES ClC1=CC=C(Br)C=C1

Specifications

Melting Point 63°C to 67°C
Density 1.651
Boiling Point 196°C
Quantity 1000 g
Odor Aromatic
UN Number UN2811
Beilstein 1099014
Solubility Information Soluble in water 0.045g/L.
Formula Weight 191.46
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-4-chlorobenzene
Show More Show Less
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-392-3
RTECSNumber CY9006000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.