Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-dodecylbenzene 90.0+%, TCI America™
SDP

Catalog No. B161225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B161225G 25 g
B16125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B161225G Supplier TCI America Supplier No. B161225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 126930-72-1
Molecular Formula C18H29Br
Molecular Weight (g/mol) 325.334
MDL Number MFCD00191386
InChI Key RFFMRBISTZHCNY-UHFFFAOYSA-N
Synonym 4-dodecylbromobenzene, 1-bromo-4-n-dodecylbenzene, benzene, 1-bromo-4-dodecyl, 4-bromododecylbenzene, pubchem22735, acmc-209bby, 1-bromo-4-dodecyl-benzene, ksc494g6d, 4-n-dodecylbromobenzene
PubChem CID 15120506
IUPAC Name 1-bromo-4-dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 191°C
Quantity 25 g
Formula Weight 325.33
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-dodecylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.