Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-iodobenzene, 98%

Catalog No. p-7021501
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 589-87-7 | C6H4BrI | 282.906 g/mol

It is mainly used as the OLED pharmaceutical intermediate, as reagent for in situ desilylation and coupling of silylated alkynes, as substrate in copper-free Sonogashira coupling in aqueous acetone. It is also used in synthesis of β,β,dibromostyrenes, as starting reagent in the total syntheses of ent-conduramine A and ent-7-deoxypancratistatin (alkaloids).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used as the OLED pharmaceutical intermediate, as reagent for in situ desilylation and coupling of silylated alkynes, as substrate in copper-free Sonogashira coupling in aqueous acetone. It is also used in synthesis of β,β,dibromostyrenes, as starting reagent in the total syntheses of ent-conduramine A and ent-7-deoxypancratistatin (alkaloids).

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-87-7
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.906
MDL Number MFCD00001051
InChI Key UCCUXODGPMAHRL-UHFFFAOYSA-N
Synonym 4-bromoiodobenzene, benzene, 1-bromo-4-iodo, p-bromoiodobenzene, p-iodobromobenzene, p-bromophenyl iodide, 4-bromo-1-iodobenzene, 1-bromo-4-iodo-benzene, 4-iodobromobenzene, rarechem fh 1g 0a12, p-bromo iodobenzene
PubChem CID 11522
IUPAC Name 1-bromo-4-iodobenzene
SMILES C1=CC(=CC=C1Br)I

Specifications

Melting Point 89°C to 93°C
Boiling Point 252°C
Odor Recognizable
Assay Percent Range 98%
Quantity 10 g
Beilstein 1904545
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 282.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-iodobenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-662-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.