Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-propoxybenzene 98.0+%, TCI America™
SDP

Catalog No. B363925G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 39969-56-7
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00156160
InChI Key VVPARGBRVKRZJC-UHFFFAOYSA-N
Synonym 4-n-propoxybromobenzene, 1-bromo-4-propoxylbenzene, 1-bromo-4-n-propoxybenzene, 1-bromo-4-n-propyloxybenzene, p-bromphenyl-n-propylather, acmc-209j9h, 1-bromo-4-propoxy-benzene, 1-4-bromophenoxy propane, 4-bromophenyl propyl ether, 1-bromanyl-4-propoxy-benzene
PubChem CID 2734198
IUPAC Name 1-bromo-4-propoxybenzene
SMILES CCCOC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 g
Formula Weight 215.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-propoxybenzene
EINECSNumber (3)-0845
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.