Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromobenzyl methyl ether, 98%

Catalog No. p-3593218
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 52711-30-5 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52711-30-5
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD03790887
InChI Key QFAZLCRHLRJNAW-UHFFFAOYSA-N
Synonym 1-bromo-2-methoxymethyl benzene, 2-bromobenzyl methyl ether, acmc-20ag76, methyl 2-bromobenzyl ether, 1-methoxymethyl-2-bromobenzene, 2-bromobenzyl alcohol, methyl ether, benzene,1-bromo-2-methoxymethyl
PubChem CID 12927162
IUPAC Name 1-bromo-2-(methoxymethyl)benzene
SMILES COCC1=CC=CC=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5470 to 1.5490 (20°C, 589nm)
Linear Formula BrC6H4CH2OCH3
Quantity 1 g
Formula Weight 201.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromobenzyl methyl ether
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.