Learn More
1-Bromobutane, 98+%
Description
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.
Solubility
Miscible with ethanol and diethyl ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, potassium, sodium and sodium oxides.
Specifications
Specifications
| Melting Point | -112°C |
| Density | 1.275 |
| Boiling Point | 101°C to 102°C |
| Flash Point | 20°C (68°F) |
| Odor | Characteristic |
| Assay Percent Range | >98% |
| Refractive Index | 1.44 |
| Linear Formula | CH3(CH2)3Br |
| Quantity | 10,000 g |
| UN Number | UN1126 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-BROMOBUTANE |
| EINECSNumber | 203-691-9 |
| RTECSNumber | EJ6225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.