Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromobutane, 98+%

Catalog No. AAA106960E
Click to view available options
Quantity:
10,000 g
2500 g
500 g

CAS: 109-65-9 | C4H9Br | 137.02 g/mol

1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

Solubility
Miscible with ethanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, potassium, sodium and sodium oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-65-9
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000260
InChI Key MPPPKRYCTPRNTB-UHFFFAOYSA-N
Synonym butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831
PubChem CID 8002
IUPAC Name 1-bromobutane
SMILES CCCCBr

Specifications

Melting Point -112°C
Density 1.275
Boiling Point 101°C to 102°C
Flash Point 20°C (68°F)
Odor Characteristic
Assay Percent Range >98%
Refractive Index 1.44
Linear Formula CH3(CH2)3Br
Quantity 2500 g
UN Number UN1126
Beilstein 1098260
Merck Index 14,1553
Solubility Information Miscible with ethanol and diethyl ether. Immiscible with water.
Formula Weight 137.03
Percent Purity ≥98%
Chemical Name or Material 1-Bromobutane
Show More Show Less
Hazard Category H225-H315-H319
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-BROMOBUTANE
EINECSNumber 203-691-9
RTECSNumber EJ6225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.