Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromobutane, 98+%

Manufacturer:  Alfa Aesar A1069636

Quantity 500g

 View more versions of this product

Catalog No. AAA1069636


  Check Availability
Add to cart

Specifications

Description

Applications
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

Solubility
Miscible with ethanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, potassium, sodium and sodium oxides.
Description & Specifications

Chemical Identifiers

109-65-9
CH3(CH2)3Br
MFCD00000260
1098260
n-Butyl bromide
C4H9Br
137.03
UN1126
14,1553
1-Bromobutane

Specifications

109-65-9
500g
C4H9Br
137.03
UN1126
14,1553
1-Bromobutane
1.275g/mL
>98%
CH3(CH2)3Br
MFCD00000260
1098260
n-Butyl bromide
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H315-H319-H335

GHS P Statement
P210-P273-P280g-P305+P351+P338

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-BROMOBUTANE

RTECSNumber : EJ6225000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit