Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromocarbazole 98.0+%, TCI America™
SDP

Catalog No. B48161G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 16807-11-7
Molecular Formula C12H8BrN
Molecular Weight (g/mol) 246.107
MDL Number MFCD18450164
InChI Key VCDOOGZTWDOHEB-UHFFFAOYSA-N
PubChem CID 11230335
IUPAC Name 1-bromo-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C(=CC=C3)Br

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 246.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromocarbazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.