Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromodocosane 98.0+%, TCI America™
SDP

Catalog No. B0392250G
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 6938-66-5
Molecular Formula C22H45Br
Molecular Weight (g/mol) 389.506
MDL Number MFCD00013543
InChI Key QYOXLKAKUAASNA-UHFFFAOYSA-N
Synonym Docosyl Bromide
PubChem CID 81355
IUPAC Name 1-bromodocosane
SMILES CCCCCCCCCCCCCCCCCCCCCCBr

Specifications

Melting Point 44°C
Color White-Yellow
Quantity 250 g
Formula Weight 389.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromodocosane
EINECSNumber (2)-0067
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.