Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromododecane, 98%

Catalog No. p-7080869
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 143-15-7 | C12H25Br | 249.236 g/mol

1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-15-7
Molecular Formula C12H25Br
Molecular Weight (g/mol) 249.236
MDL Number MFCD00000225
InChI Key PBLNBZIONSLZBU-UHFFFAOYSA-N
Synonym dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
PubChem CID 8919
IUPAC Name 1-bromododecane
SMILES CCCCCCCCCCCCBr

Specifications

Melting Point -10°C
Density 1.04
Boiling Point 276°C
Flash Point 146°C (294°F)
Odor Weak
Refractive Index 1.458
Linear Formula CH3(CH2)11Br
Quantity 250 g
Beilstein 506159
Merck Index 14,5389
Solubility Information Insoluble in water.
Formula Weight 249.24
Percent Purity 98%
Chemical Name or Material 1-Bromododecane
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-587-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.