Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromoheptadecane 98.0+%, TCI America™
SDP

Catalog No. B093725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B093725G 25 g
B09375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B093725G Supplier TCI America Supplier No. B093725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3508-00-7
Molecular Formula C17H35Br
Molecular Weight (g/mol) 319.371
MDL Number MFCD00017891
InChI Key HHSDZLLPIXMEIU-UHFFFAOYSA-N
Synonym Heptadecyl Bromide
PubChem CID 19022
IUPAC Name 1-bromoheptadecane
SMILES CCCCCCCCCCCCCCCCCBr

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 319.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromoheptadecane
EINECSNumber (2)-0067
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.