Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromohexane, 99%

Catalog No. aaa1516636
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

Solubility
Immiscible with water.

Notes
Keep away from heat and sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-25-1
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.07
MDL Number MFCD00000271
InChI Key MNDIARAMWBIKFW-UHFFFAOYSA-N
Synonym hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
PubChem CID 8101
IUPAC Name 1-bromohexane
SMILES CCCCCCBr

Specifications

Melting Point -85°C
Density 1.176
Boiling Point 156°C
Flash Point 50°C (122°F)
Odor Pleasant
Refractive Index 1.4475
Quantity 500 g
UN Number UN1993
Beilstein 1731290
Solubility Information Immiscible with water.
Formula Weight 165.08
Percent Purity 99%
Chemical Name or Material 1-Bromohexane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315
Flammable liquid and vapour.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-850-2
RTECSNumber MO0925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.