Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromonaphthalene, 96%

Catalog No. AC107150025
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107150025 2.5 L Glass bottle
AC107151000 100 mL Glass bottle
AC107155000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107150025 Supplier Thermo Scientific Chemicals Supplier No. 107150025
Add to Cart

CAS: 90-11-9 | C10H7Br | 207.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-11-9 , 580-13-2
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Decomposition Information 180°C
Melting Point -1.0°C
Color Brown-Yellow to Yellow
Density 1.4890g/mL
Boiling Point 279.0°C to 281.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 2.5% max. 2-Bromonaphthalene (GC)
Packaging Glass bottle
Molecular Formula C10H7Br
Refractive Index 1.6560 to 1.6590
Linear Formula C10H7Br
Quantity 2.5 L
Beilstein 05, 547
Specific Gravity 1.489
Merck Index 15, 1436
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether and ethanol
Vapor Pressure 14.8hPa at 20°C
Formula Weight 207.07
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-965-2
RTECSNumber QJ1545000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.