Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromonaphthalene 95.0+%, TCI America™
SDP

Catalog No. B0618100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0618100G 100 g
B061825G 25 g
B0618500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0618100G Supplier TCI America Supplier No. B0618100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90-11-9
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 282°C
Quantity 100 g
Formula Weight 207.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene
RTECSNumber QJ1545000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.