Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromopentane, 99%

Catalog No. AAA108210B
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 110-53-2 | C5H11Br | 151.047 g/mol

1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

Solubility
Insoluble in water and soluble in alcohol. Miscible in diethyl ether.

Notes
Store away from oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-53-2
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.047
MDL Number MFCD00000267
InChI Key YZWKKMVJZFACSU-UHFFFAOYSA-N
Synonym n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0
PubChem CID 8057
IUPAC Name 1-bromopentane
SMILES CCCCCBr

Specifications

Melting Point -95°C
Density 1.215
Boiling Point 127°C to 129°C
Flash Point 31°C (87°F)
Odor Ether-like
Refractive Index 1.4436
Quantity 1000 g
UN Number UN1993
Beilstein 1730981
Merck Index 14,602
Solubility Information Insoluble in water and soluble in alcohol. Miscible in diethyl ether.
Formula Weight 151.05
Percent Purity 99%
Chemical Name or Material 1-Bromopentane
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-776-0
RTECSNumber RZ9770000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.