Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromopyrene 95.0+%, TCI America™
SDP

Catalog No. B149525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B149525G 25 g
B14955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B149525G Supplier TCI America Supplier No. B149525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1714-29-0
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.152
MDL Number MFCD00015767
InChI Key HYGLETVERPVXOS-UHFFFAOYSA-N
Synonym 1-bromo-pyrene, pyrene, bromo, pyrene, 1-bromo, bromopyrene, 1-brompyren, 1-brpy, 3-bromopyrene, 1-bromanylpyrene, pubchem14567, 1-bromopyrene
PubChem CID 159627
IUPAC Name 1-bromopyrene
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)Br

Specifications

Melting Point 98°C
Color White-Yellow
Boiling Point 255°C
Quantity 25 g
Formula Weight 281.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromopyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.