Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanethiol, 98%

Catalog No. AC150802500
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 109-79-5 | C4H10S | 90.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-79-5
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00004905
InChI Key WQAQPCDUOCURKW-UHFFFAOYSA-N
Synonym 1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
PubChem CID 8012
IUPAC Name butane-1-thiol
SMILES CCCCS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -116.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 98.0°C
Flash Point 0.5°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4420 to 1.4440
Linear Formula CH3(CH2)3SH
Quantity 250 mL
Beilstein 01, 370
Fieser 02,53; 04,64
Specific Gravity 0.84
Merck Index 15, 1580
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 90.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Butanethiol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 203-705-3
RTECSNumber EK6300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.