Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, ACS, 99.4+%, Thermo Scientific Chemicals

Catalog Number AA31068AP
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 x 4 L
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.005%, Titratable acid 0.0008meq/g, Carbonyl compounds (as butyraldehyde) 0.01%, Butyl ether 0.2%, H2O 0.1%

1-Butanol is used as solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Appearance Colorless liquid
Density 0.810 g/cm3 at 20°C
Boiling Point 117°C to 118°C
Quantity 500 mL
Flash Point 35°C (95°F)
Odor Alcool-like
Physical Form Liquid
Refractive Index 1.399
Assay Percent Range >99.4%
Vapor Pressure 5.6 hPa (4mm Hg) at 20°C
Beilstein 969148
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water.
Formula Weight 74.12
Grade ACS
Concentration or Composition (by Analyte or Components) 1-Butanol
Chemical Name or Material 1-Butanol
Show More Show Less
Hazard Category H226-H302-H315-H318-H335-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: III
EINECSNumber 200-751-6
RTECSNumber EO1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit