Learn More
1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals ProductsDescription
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Specifications
Specifications
| Melting Point | -90°C |
| Density | 0.81g/mL at 25°C |
| Boiling Point | 116°C to 118°C |
| Quantity | 4 x 2.5 L |
| Refractive Index | n20/D 1.399 |
| Linear Formula | CH3(CH2)3OH |
| MDL Number | MFCD00002964 |
| Beilstein | 969148 |
| Synonym | 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane |
| Molecular Weight (g/mol) | 74.123 |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235 |
| Precautionary Statement | H226-H302-H315-H318-H335-H336 |
| EINECSNumber | 200-751-6 |
| RTECSNumber | EO1400000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.