Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%

Catalog No. p-7030267
Click to view available options
Quantity:
5 g
50 g

Catalog No. AAH2782706

Supplier: Thermo Scientific Chemicals H2782706

Only null left
Add to Cart

CAS: 227617-70-1 | C9H17F6N2P | 298.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 227617-70-1
Molecular Formula C9H17F6N2P
Molecular Weight (g/mol) 298.21
MDL Number MFCD03790877
InChI Key JWFPQAXAGSAKRF-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636
PubChem CID 11243457
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Melting Point 48°C to 50°C
Density 1.354
Refractive Index 1.423
Quantity 5 g
Formula Weight 298.21
Percent Purity 99%
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.