Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. B247525G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 402846-78-0
Molecular Formula C9H17BF4N2
Molecular Weight (g/mol) 240.05
MDL Number MFCD03427618
InChI Key VCAIYEJBOWHUGP-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b
PubChem CID 2734244
IUPAC Name 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 240.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.