Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
174501-64-5
C8H15N2·PF6
284.18
IXQYBUDWDLYNMA-UHFFFAOYSA-N
1-butyl-3-methylimidazolium hexafluorophosphate,1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v,unii-zge3n4o8q9,1-butyl-3-methylimidazoliumhexafluorophosphate,1-n-butyl-3-methylimidazolium hexafluorophosphate,zge3n4o8q9,butylmethylimidazolium hexafluorophosphate,1-methyl-3-butylimidazolium hexafluorophosphate,bmim pf6,c4mim hexafluorophosphate
2734174
1-butyl-3-methylimidazol-3-ium;hexafluorophosphate
CCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F
174501-64-5
C8H15N2·PF6
284.18
IXQYBUDWDLYNMA-UHFFFAOYSA-N
1-butyl-3-methylimidazolium hexafluorophosphate,1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v,unii-zge3n4o8q9,1-butyl-3-methylimidazoliumhexafluorophosphate,1-n-butyl-3-methylimidazolium hexafluorophosphate,zge3n4o8q9,butylmethylimidazolium hexafluorophosphate,1-methyl-3-butylimidazolium hexafluorophosphate,bmim pf6,c4mim hexafluorophosphate
2734174
1-butyl-3-methylimidazol-3-ium;hexafluorophosphate
CCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F
Ionic liquid
Specific Gravity 1.3727
Packaging Glass bottle
Boiling Point >340.0°C
Infrared Spectrum Authentic
Quantity 100g
Density 1.3727g/mL
Melting Point 6.5°C
Color Colorless to Yellow
Water 1% Max. (K.F.)
Viscosity 272.1 mPa.s (25°C)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Merck Index 15,1584
Formula Weight 284.18
Physical Form Viscous Liquid
Percent Purity ≥98%
Chemical Name or Material 1-Butyl-3-methylimidazolium hexafluorophosphate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning