Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butylpiperazine 98.0+%, TCI America™
SDP

Catalog No. B407825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B407825ML 25 mL
B40785ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B407825ML Supplier TCI America Supplier No. B407825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5610-49-1
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00039702
InChI Key YKSVXVKIYYQWBB-UHFFFAOYSA-N
PubChem CID 424322
IUPAC Name 1-butylpiperazine
SMILES CCCCN1CCNCC1

Specifications

Color Yellow
Boiling Point 192°C
Quantity 25 mL
Formula Weight 142.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.