Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butylpyridinium Tetrafluoroborate 94.0+%, TCI America™
SDP

Catalog No. B323225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B323225G 25 g
B32325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B323225G Supplier TCI America Supplier No. B323225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 203389-28-0
Molecular Formula C9H14BF4N
Molecular Weight (g/mol) 223.022
MDL Number MFCD03427602
InChI Key XLWDQAHXRCBPEE-UHFFFAOYSA-N
Synonym 1-butylpyridinium tetrafluoroborate, 1-butylpyridin-1-ium tetrafluoroborate, n-butylpyridinium tetrafluoroborate, bpmbf4, acmc-1co7o, dsstox_cid_29097, dsstox_rid_83316, dsstox_gsid_49241, ksc199a6h, 1-butylpyridin-1-ium; tetrafluoroboranuide
PubChem CID 2734180
IUPAC Name 1-butylpyridin-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CCCC[N+]1=CC=CC=C1

Specifications

Color Yellow
Quantity 25 g
UN Number 1760
Formula Weight 223.02
Percent Purity ≥94.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Butylpyridinium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.