Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Carbobenzoxy-3-pyrrolidone 98.0+%, TCI America™
SDP

Catalog No. C24371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24371G 1 g
C24375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24371G Supplier TCI America Supplier No. C24371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130312-02-6
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD03001711
InChI Key LMHWEUQNJRXMCD-UHFFFAOYSA-N
Synonym 1-n-cbz-3-pyrrolidinone, 1-cbz-3-pyrrolidinone, n-cbz-3-pyrrolidinone, 1-cbz-3-pyrrolidone, 1-benzyloxycarbonyl-3-pyrrolidinone, 1-z-3-pyrrolidinone, n-benzyloxycarbonyl-3-pyrrolidinone, 1-carbobenzoxy-3-pyrrolidone, 1-pyrrolidinecarboxylic acid, 3-oxo-, phenylmethyl ester, 3-oxo-pyrrolidine-1-carboxylic acid benzyl ester
PubChem CID 561203
IUPAC Name benzyl 3-oxopyrrolidine-1-carboxylate
SMILES C1CN(CC1=O)C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 46°C
Quantity 1 g
Formula Weight 219.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Carbobenzoxy-3-pyrrolidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.