Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers


Click to view available options
Volume (Metric):
1 nL to 200 μL
Channels:
1
Description
Great choice for dispensing picoliter to microliter volumes for one to eight different types of fluid across an entire plate or just part of a plate.
- Cost Saving through assay miniaturization
- Improved efficiency and sample integrity
- Powerful, flexible, and easy to use software

Specifications
Specifications
Product Type | Digital Dispenser Cartridge |
Autoclavable | Not Autoclavable |
Channels | 1 |
Volume (Metric) | 1 nL to 200 μL |
For Use With (Application) | Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting |
For Use With (Equipment) | Mulitdrop Pico 1 Digital Dispenser |
Interface | USB |
Plate Type | 12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm |
Precision | ≤ 8% |
Software | PicoIT 1 |
Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction