Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-(chloromethyl)-3,5-dioxahexane 95.0+%, TCI America™
SDP

Catalog No. C119725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C119725G 25 g
C11975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C119725G Supplier TCI America Supplier No. C119725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70905-45-2
Molecular Formula C5H10Cl2O2
Molecular Weight (g/mol) 173.033
MDL Number MFCD00130114
InChI Key GEGJQMIXBNYSQG-UHFFFAOYSA-N
Synonym 1,1-Bis(chloromethyl)-2,4-dioxapentane, 1,3-Dichloro-2-(methoxymethoxy)propane
PubChem CID 5323878
IUPAC Name 1,3-dichloro-2-(methoxymethoxy)propane
SMILES COCOC(CCl)CCl

Specifications

Color Colorless
Boiling Point 95°C
Quantity 25 g
Formula Weight 173.03
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-(chloromethyl)-3,5-dioxahexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.