Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-nitrobenzene, 99+%

Catalog No. p-89392
Click to view available options
Quantity:
100 mL
1 L

CAS: 88-73-3 | C6H4ClNO2 | 157.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-73-3
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00007061
InChI Key BFCFYVKQTRLZHA-UHFFFAOYSA-N
Synonym 2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
PubChem CID 6945
ChEBI CHEBI:34270
IUPAC Name 1-chloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 32°C to 34°C
Color Yellow
Density 1.3480g/mL
Boiling Point 246°C
Flash Point 124°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula ClC6H4NO2
Quantity 100 mL
Beilstein 05, 241
Specific Gravity 1.348
Merck Index 15, 2153
Solubility Information Solubility in water: 0.43g/L water (20°C).,soluble in alcohol,benzene,ether,acetone,carbo,toluene,pyridine
Formula Weight 157.56
Percent Purity 99+%
Physical Form Fused Crystalline Mass
Chemical Name or Material 1-Chloro-2-nitrobenzene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-854-9
RTECSNumber CZ0875000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.