Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-octyne, 98%, Thermo Scientific Chemicals

Catalog No. AAH5345206
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51575-83-8
Molecular Formula C8H13Cl
Molecular Weight (g/mol) 144.642
MDL Number MFCD00274223
InChI Key OUMUOQQKEAGFCJ-UHFFFAOYSA-N
Synonym 1-chloro-2-octyne, acmc-1asda, oumuoqqkeagfcj-uhfffaoysa
PubChem CID 4349444
IUPAC Name 1-chlorooct-2-yne
SMILES CCCCCC#CCCl

Specifications

Density 0.931
Boiling Point 81°C (15 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.46
Linear Formula CH3(CH2)4C≡CCH2Cl
Quantity 5 g
Formula Weight 144.64
Percent Purity 98%
Chemical Name or Material 1-Chloro-2-octyne
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.