Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-propanol, tech. 75% (remainder mainly 2-chloro-1-propanol)

Catalog No. aab2268614
Click to view available options
Quantity:
25 g
100 g

CAS: 127-00-4 | C3H7ClO | 94.538 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-00-4
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.538
MDL Number MFCD00004530
InChI Key YYTSGNJTASLUOY-UHFFFAOYSA-N
Synonym 1-chloro-2-propanol, 2-propanol, 1-chloro, chloroisopropyl alcohol, propene chlorohydrin, 1-chloroisopropyl alcohol, sec-propylene chlorohydrin, 1-chloro-2-hydroxypropane, .alpha.-propylene chlorohydrin, ccris 4766, 3-chloro-2-propanol
PubChem CID 31370
ChEBI CHEBI:76260
IUPAC Name 1-chloropropan-2-ol
SMILES CC(CCl)O

Specifications

Density 1.111
Boiling Point 126°C to 127°C
Flash Point 51°C (123°F)
Refractive Index 1.437
Quantity 25 g
UN Number UN2929
Beilstein 773653
Merck Index 14,7850
Formula Weight 94.54
Percent Purity 75%
Grade Technical
Assay remainder mainly 2-chloro-1-propanol
Chemical Name or Material 1-Chloro-2-propanol
Show More Show Less
Hazard Category H226-H301+H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H301-H311-H330-H226-H341-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 204-819-6
RTECSNumber UA8942000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.