Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3,4-dinitrobenzene, 90%, Thermo Scientific™

Catalog No. p-4229139
Click to view available options
Quantity:
50 g
100 g
250 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 610-40-2
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
InChI Key QVQSOXMXXFZAKU-UHFFFAOYSA-N
Synonym 1-chloro-3,4-dinitrobenzene, 3,4-dinitrochlorobenzene, benzene, 4-chloro-1,2-dinitro, unii-z29b1f274e, 1,2-dinitro-4-chlorobenzene, 1,2-dino2 4-cl benzene, 4-chlordinitrobenzol, acmc-209mog, 4-chloro-1,2-dinitrobenzen, ghl.pd_mitscher_leg0.928
PubChem CID 33097
IUPAC Name 4-chloro-1,2-dinitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 36.0°C
Color Brown-Yellow
Density 1.6800g/mL
Boiling Point 315.0°C
Flash Point 194°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5865 to 1.5885 (20°C, 589nm)
Linear Formula ClC6H3(NO2)2
Quantity 50 g
Beilstein 05,262
Specific Gravity 1.68
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 202.55
Percent Purity 90%
Physical Form Oily Liquid After Melting
Chemical Name or Material 1-Chloro-3,4-dinitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 210-223-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.