Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-fluoro-2-propanol, 95%

Catalog No. AAL1728606
Click to view available options
Quantity:
5 g

CAS: 453-11-2 | C3H6ClFO | 112.528 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 453-11-2
Molecular Formula C3H6ClFO
Molecular Weight (g/mol) 112.528
MDL Number MFCD00045284
InChI Key ZNKJYZHZWALQNX-UHFFFAOYSA-N
Synonym 1-chloro-3-fluoroisopropanol, 1-chloro-3-fluoro-2-propanol, 2-propanol, 1-chloro-3-fluoro, wln: g1yq1f, sgqdlhbrbrwup@, glycerolchlorofluorohydrin, 1-chloro-3-fluoro-propan-2-ol, 5-bromo-2,4-difluorobenzenamine, 1-fluoro-3-chloro isopropanol
PubChem CID 92107
IUPAC Name 1-chloro-3-fluoropropan-2-ol
SMILES C(C(CCl)O)F

Specifications

Density 1.3
Boiling Point 153°C to 156°C
Refractive Index 1.428
Quantity 5 g
UN Number UN1992
Beilstein 1732501
Formula Weight 112.53
Percent Purity 95%
Chemical Name or Material 1-Chloro-3-fluoro-2-propanol
Hazard Category H226-H302+H312+H332
Hazard Statement GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.