Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-nitrobenzene, 98%

Catalog No. p-7027001
Click to view available options
Quantity:
250 g

CAS: 121-73-3 | C6H4ClNO2 | 157.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-73-3
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00007202
InChI Key KMAQZIILEGKYQZ-UHFFFAOYSA-N
Synonym m-chloronitrobenzene, 3-nitrochlorobenzene, 3-chloronitrobenzene, m-nitrochlorobenzene, benzene, 1-chloro-3-nitro, chloro-m-nitrobenzene, 3-chloro-1-nitrobenzene, metachloronitrobenzene, 1-chloronitrobenzene, 1-nitro-3-chlorobenzene
PubChem CID 8489
ChEBI CHEBI:82420
IUPAC Name 1-chloro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 43°C to 47°C
Density 1.34
Boiling Point 235°C to 236°C
Flash Point 124°C (255°F)
Quantity 250 g
UN Number UN1578
Beilstein 1907947
Merck Index 14,2151
Formula Weight 157.56
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-nitrobenzene
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H311-H373-H302
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORONITROBENZENES, SOLID
EINECSNumber 204-496-1
RTECSNumber CZ0940000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.