Learn More
1-Chloro-4-nitrobenzene, 98+%
Description
1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.
Solubility
Soluble in boiling hot ethanol, ether, toluene, acetone, organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 82°C to 86°C |
| Density | 1.515 |
| Boiling Point | 238°C to 242°C |
| Flash Point | 127°C (260°F) |
| Quantity | 250 g |
| UN Number | UN1578 |
| Beilstein | 508691 |
| Merck Index | 14,2151 |
| Solubility Information | Soluble in boiling hot ethanol,ether,toluene,acetone,organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water. |
| Formula Weight | 157.56 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331-H341-H351-H373-H500 |
| Hazard Statement | GHS H Statement H301-H311-H331-H341-H351-H373 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Suspected of causing genetic defects. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORONITROBENZENES, SOLID |
| EINECSNumber | 202-809-6 |
| RTECSNumber | CZ1050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.