Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-5-triethylsilyl-4-pentyne, 97%

Catalog No. AAH5351514
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH5351514

Supplier: Thermo Scientific Chemicals H5351514

Only null left
Add to Cart

CAS: 174125-30-5 | C11H21ClSi | 216.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174125-30-5
Molecular Formula C11H21ClSi
Molecular Weight (g/mol) 216.82
MDL Number MFCD00671352
InChI Key YOFYXFCOGXKCGG-UHFFFAOYSA-N
Synonym 1-chloro-5-triethylsilyl-4-pentyne, 5-chloropent-1-ynyl triethyl silane, 5-chloropent-1-ynyl triethylsilane, 1-triethylsilyl-5-chloro-1-pentyne, 5-chloranylpent-1-ynyl triethyl silane, 5-chloropent-1-yn-1-yl triethylsilane, 5-chloropent-1-yn-1-yl triethyl silane, silane, 5-chloro-1-pentyn-1-yl triethyl
PubChem CID 2757930
IUPAC Name 5-chloropent-1-ynyl(triethyl)silane
SMILES CC[Si](CC)(CC)C#CCCCCl

Specifications

Quantity 25 g
Linear Formula Cl(CH2)3C≡CSi(CH2CH3)3
Formula Weight 216.83
Percent Purity 97%
Chemical Name or Material 1-Chloro-5-triethylsilyl-4-pentyne
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.