Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorodocosane 98.0+%, TCI America™
SDP

Catalog No. C077525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C077525G 25 g
C07755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C077525G Supplier TCI America Supplier No. C077525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42217-03-8
Molecular Formula C22H45Cl
Molecular Weight (g/mol) 345.05
MDL Number MFCD00039383
InChI Key OACXFSZVCDOBKF-UHFFFAOYSA-N
Synonym Docosyl Chloride
PubChem CID 545602
IUPAC Name 1-chlorodocosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCl

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 25 g
Formula Weight 345.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chlorodocosane
EINECSNumber (2)-0066&(2)-0068
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.