Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloroethyl Ethyl Carbonate 98.0+%, TCI America™
SDP

Catalog No. C1056100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1056100G 100 g
C105625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1056100G Supplier TCI America Supplier No. C1056100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50893-36-2
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.574
MDL Number MFCD00040523
InChI Key YVRGKFXJZCTTRB-UHFFFAOYSA-N
Synonym 1-chlorodiethyl carbonate, carbonic acid, 1-chloroethyl ethyl ester, 1-chloroethyl ethylcarbonate, 1-ethoxycarbonyloxyethyl chloride, carbonic acid 1-chloroethyl ethyl ester, 1-chloroethylethylcarbonate, acmc-1aw9h, 1-chloroethyl ethoxyformate, ethyl 1-chloroethyl carbonate, ksc490c2n
PubChem CID 162095
IUPAC Name 1-chloroethyl ethyl carbonate
SMILES CCOC(=O)OC(C)Cl

Specifications

Color Yellow
Boiling Point 160°C
Quantity 100 g
Formula Weight 152.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloroethyl Ethyl Carbonate
EINECSNumber (2)-3303
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.