Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chlorohexadecane, 97%

Click to view available options
Quantity:
100mL
500mL
4864-01-1
C9H7F3O3
220.15
MFCD00000959
NAKZCKOHULJEID-UHFFFAOYSA-N
2-methoxy-5-trifluoromethyl benzoic acid,benzoic acid, 2-methoxy-5-trifluoromethyl,pubchem10415,acmc-209wn0,benzoic acid,2-methoxy-5-trifluoromethyl,2-methyloxy-5-trifluoromethyl benzoic acid,2-methoxy-5-trifluoromethyl benzoicacid
13585909
2-methoxy-5-(trifluoromethyl)benzoic acid
COC1=C(C=C(C=C1)C(F)(F)F)C(=O)O
4864-01-1
C9H7F3O3
220.15
MFCD00000959
NAKZCKOHULJEID-UHFFFAOYSA-N
2-methoxy-5-trifluoromethyl benzoic acid,benzoic acid, 2-methoxy-5-trifluoromethyl,pubchem10415,acmc-209wn0,benzoic acid,2-methoxy-5-trifluoromethyl,2-methyloxy-5-trifluoromethyl benzoic acid,2-methoxy-5-trifluoromethyl benzoicacid
13585909
2-methoxy-5-(trifluoromethyl)benzoic acid
COC1=C(C=C(C=C1)C(F)(F)F)C(=O)O
Applications
1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

Solubility
Soluble in ether. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Quantity 100mL
Boiling Point 322°C
Flash Point 136°C (276°F)
Melting Point 8°C to 10°C
Linear Formula CH3(CH2)15Cl
Beilstein 1748498
Formula Weight 260.89
Percent Purity 97%
Chemical Name or Material 1-Chlorohexadecane