Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorohexadecane, 97%

Catalog No. p-7026989
Click to view available options
Quantity:
100 mL
500 mL

CAS: 4860-03-1 | C9H7F3O3 | 220.15 g/mol

1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

Solubility
Soluble in ether. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4860-03-1
MDL Number MFCD00000959

Specifications

Melting Point 8°C to 10°C
Density 0.865
Boiling Point 322°C
Flash Point 136°C (276°F)
Molecular Formula C9H7F3O3
Refractive Index 1.449
Linear Formula CH3(CH2)15Cl
Quantity 100 mL
Beilstein 1748498
Synonym 2-methoxy-5-trifluoromethyl benzoic acid, benzoic acid, 2-methoxy-5-trifluoromethyl, pubchem10415, acmc-209wn0, benzoic acid,2-methoxy-5-trifluoromethyl, 2-methyloxy-5-trifluoromethyl benzoic acid, 2-methoxy-5-trifluoromethyl benzoicacid
Solubility Information Soluble in ether. Insoluble in water.
InChI Key NAKZCKOHULJEID-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)C(F)(F)F)C(=O)O
IUPAC Name 2-methoxy-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 220.15
PubChem CID 13585909
Formula Weight 260.89
Percent Purity 97%
Chemical Name or Material 1-Chlorohexadecane
Show More Show Less
EINECSNumber 225-461-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.