Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorohexane, 95%

Catalog No. ac404640050
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

CAS: 544-10-5 | C6H13Cl | 120.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-10-5
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00001018
InChI Key MLRVZFYXUZQSRU-UHFFFAOYSA-N
Synonym chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride
PubChem CID 10992
IUPAC Name 1-chlorohexane
SMILES CCCCCCCl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 0.8700g/mL
Boiling Point 135.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4189 to 1.4209
Linear Formula CH3(CH2)5Cl
Quantity 5 g
Beilstein 01, 143
Specific Gravity 0.87
Merck Index 15, 2145
Formula Weight 120.62
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-chlorohexane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-859-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.