Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorononane, 99%, Thermo Scientific Chemicals

Catalog No. AC218640100
Click to view available options
Quantity:
10 mL

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2473-01-0
Molecular Formula C9H19Cl
Molecular Weight (g/mol) 162.701
MDL Number MFCD00001023
InChI Key RKAMCQVGHFRILV-UHFFFAOYSA-N
Synonym nonyl chloride, nonane, 1-chloro, n-nonyl chloride, nonane,1-chloro, tetrapropenyl chloride, 1-propene, tetramer, chlorinated, acmc-209gec, 1-chlorononane, dsstox_cid_29111, dsstox_rid_83330
PubChem CID 17185
IUPAC Name 1-chlorononane
SMILES CCCCCCCCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4340 to 1.4360 (20°C, 589nm)
Linear Formula CH3(CH2)8Cl
Quantity 10 mL
Beilstein 01, II, 128
Formula Weight 162.7
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1-Chlorononane, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.